missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mannose Phosphate Isomerase Antibody (OTI4G7), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72584AF488
This item is not returnable.
View return policy
Description
Mannose Phosphate Isomerase Monoclonal specifically detects Mannose Phosphate Isomerase in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| Mannose Phosphate Isomerase | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MPI | |
| Full length recombinant protein of human Mannose Phosphate Isomerase (NP_002426) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI4G7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| CDG1B, EC 5.3.1, EC 5.3.1.8, FLJ39201, mannose phosphate isomerase, mannose-6- phosphate isomerase, mannose-6-phosphate isomerase, Phosphohexomutase, Phosphomannose isomerase, phosphomannose isomerase 1, PMI, PMI1phosphohexomutase | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 4351.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction