Learn More
Mannose Phosphate Isomerase Antibody (OTI4G7), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-72584JF549
Product Code. 18137675
  • € 751.00 / 0.10mL
Description
Mannose Phosphate Isomerase Monoclonal specifically detects Mannose Phosphate Isomerase in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mannose Phosphate Isomerase
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
MPI
Full length recombinant protein of human Mannose Phosphate Isomerase (NP_002426) produced in HEK293T cells.
0.1 mL
Primary
Human, Rat
IgG1
Western Blot, Flow Cytometry, Immunohistochemistry
OTI4G7
Western Blot, Flow Cytometry, Immunohistochemistry
CDG1B, EC 5.3.1, EC 5.3.1.8, FLJ39201, mannose phosphate isomerase, mannose-6- phosphate isomerase, mannose-6-phosphate isomerase, Phosphohexomutase, Phosphomannose isomerase, phosphomannose isomerase 1, PMI, PMI1phosphohexomutase
Mouse
Affinity Purified
Lipid and Metabolism
4351.0
Store at 4°C in the dark.