missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MANSC1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16607284
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16607284 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16607284 Supplier Invitrogen™ Supplier No. PA5100797

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MANS1.

MANSC1 gene ontology (GO) annotation includes transmembrane transporter activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MANSC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MANSC1
Gene Accession No. Q9H8J5
Gene Alias 9130403P13Rik; LOH12CR3; Loss of heterozygosity 12 chromosomal region 3 protein; MANSC domain containing 1; MANSC domain-containing protein 1; Mansc1; UNQ316/PRO361
Gene Symbols MANSC1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MANSC1(Accession Q9H8J5), corresponding to amino acid residues P321-S371.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54682, 690606
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.