missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PADI4 Antibody (OTI4H5), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73226PCP
This item is not returnable.
View return policy
Description
PADI4 Monoclonal specifically detects PADI4 in Human samples. It is validated for Flow Cytometry.
Specifications
| PADI4 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| PADI4 | |
| Full length recombinant protein of human PADI4 (NP_036519) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| OTI4H5 | |
| Flow Cytometry | |
| HL-60 PAD, PADI5PADI-H protein, PADPAD4, PDI4, PDI5EC 3.5.3.15, peptidyl arginine deiminase, type IV, peptidyl arginine deiminase, type V, Peptidylarginine deiminase IV, Protein-arginine deiminase type IV, protein-arginine deiminase type-4 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 23569.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction