Learn More
Hanna Instruments™ Manual Calibration Dissolved Oxygen Meter
Product Code. 10248660
Click to view available options
Weight (Metric):
300 g
Unit Size:
Each
Product Code. 10248660
Brand: Hanna Instruments™ HI9142
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Includes HI76407 DO probe, HI76407A/P pre-formed PTFE DO membranes (2), HI7041S electrolyte solution (30 mL), batteries, and carrying case
Range (Polarographic DO) 0.0 to 19.9 mg/L (ppm)
Resolution (Polarographic DO) 0.1 mg/L
Accuracy (Polarographic DO) ±1.5% FS
Temperature (Metric) Range -5.0°C to +50.0°C
Resolution (Temperature) 0.1°C (1°F)
Accuracy (Temperature) ±0.2°C (±1°F) excluding probe error
Dimensions (L x W x H) 185 x 72 x 36 mm (7.3 x 2.8 x 1.4 in.)
Weight (Metric) 300 g
Weight (English) 10.6 oz.
Warranty 2 years
Power Supply 1.5 V AA batteries (4)
Probe Type Polarographic DO probe, internal temperature sensor, DIN connector
Temperature Compensation Automatic, 0°C to 50°C (32°F to 122°F)
Battery Life approx. 500 hours of use