missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAOA Recombinant Rabbit Monoclonal Antibody (4C11)
GREENER_CHOICE

Product Code. 17939052
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17939052 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17939052 Supplier Invitrogen™ Supplier No. MA549951

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

MAOA Recombinant Monoclonal Antibody for ICC/IF, IHC (P), ELISA

This gene encodes monoamine oxidase A, an enzyme that degrades amine neurotransmitters, such as dopamine, norepinephrine, and serotonin. The protein localizes to the mitochondrial outer membrane. The gene is adjacent to a related gene on the opposite strand of chromosome X. Mutation in this gene results in monoamine oxidase deficiency, or Brunner syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAOA
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Monoclonal
Clone 4C11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAOA
Gene Accession No. P21397
Gene Alias 1110061B18Rik; AA407771; Amine oxidase [flavin-containing] A; HGNC:6833; Mao; MAOA; MAO-A; monoamine oxidase A; monoamine oxidase type A; RP1-201D17__B.2
Gene Symbols MAOA
Host Species Rabbit
Immunogen A synthesized peptide derived from human Monoamine Oxidase A.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4128
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.