missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAP1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5112255

Product Code. 17170114

  • € 467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MAP1 Polyclonal Antibody for Western Blot, IHC (P)

Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. Members of the Bcl-2 family are known to be critical regulators of this process. These proteins are characterized by the presence of several conserved motifs termed Bcl-2 homology (BH) domains. A related protein termed MAP-1 has recently been identified. This protein contains a BH3-like domain and induces caspase-dependent apoptosis in mammalian cells when overexpressed. It forms homodimers and associates with Bcl-2 family members such as Bax, Bcl-2, and Bcl-XL in vitro and in vivo. It has been suggested that MAP-1 associates with the tumor suppressor RASSF1A following death receptor activation, allowing a conformational change in Bax that leads to cellular apoptosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAP1
Polyclonal
Unconjugated
MOAP1
AA987038; MAP1; Map-1; Moap1; Modulator of apoptosis 1; paraneoplastic antigen like 4; paraneoplastic antigen Ma4; paraneoplastic Ma antigen family member 4; PNMA4
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
64112
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.03% ProClin 300; pH 7
Q96BY2
MOAP1
E. coli-derived Human MAP1/MOAP1 fragment.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.