missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP1LC3A Monoclonal Antibody (OTI2D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803407S
This item is not returnable.
View return policy
Description
MAP1LC3A Monoclonal antibody specifically detects MAP1LC3A in Human samples. It is validated for Western BlotSpecifications
| MAP1LC3A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HBT8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57335 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2D5 | |
| Unconjugated | |
| ZNF286A | |
| 1010001H21Rik; 4922501H04Rik; ATG8E; Autophagy-related protein LC3 A; autophagy-related ubiquitin-like modifier LC3 A; LC3; LC3a; MAP1 light chain 3-like protein 1; MAP1A/1B light chain 3 A; MAP1A/MAP1B LC3 A; MAP1A/MAP1B light chain 3 A; MAP1ALC3; MAP1BLC3; map1lc3a; microtubule associated protein 1 light chain 3 alpha; Microtubule-asso; microtubule-associated protein 1 light chain 3 alpha; microtubule-associated protein 1-light chain 3A; microtubule-associated proteins 1A/1B light chain 3; microtubule-associated proteins 1A/1B light chain 3A; MLP3A; RP11-346K17.1; similar to microtubule-associated protein 1 light chain 3 alpha; zgc:77094 | |
| Human recombinant protein fragment corresponding to amino acids 1-252 of human ZNF286A produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction