missing translation for 'onlineSavingsMsg'
Learn More

MAP1LC3A, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit Polyclonal Antibody

Brand:  Abnova PAB31392.100uL

Product Code. 16138650

  • € 503.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: MPSDRPFKQRRSFADRCKEVQQIRDQHPSKIPVIIERYKGEKQLPVLDKTKFLVPDHVNMS
Specifications

Specifications

MAP1LC3A
Polyclonal
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:500) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user.
microtubule-associated protein 1 light chain 3 alpha
MAP1LC3A
Recombinant protein corresponding to human MAP1LC3A.
RUO
84557
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Liquid
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
LC3/LC3A/MAP1ALC3/MAP1BLC3
Rabbit
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.