missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals MAP1S Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00055201-B01P
This item is not returnable.
View return policy
Description
MAP1S Polyclonal antibody specifically detects MAP1S in Human samples. It is validated for Western Blot
Specifications
| MAP1S | |
| Polyclonal | |
| PBS (pH 7.4) | |
| BPY2 interacting protein 1, BPY2IP1VCY2IP-1, C19orf5, chromosome 19 open reading frame 5, FLJ10669, MAP8MAP-1S, MGC133087, microtubule-associated protein 1S, Microtubule-associated protein 8, Variable charge Y chromosome 2-interacting protein 1, VCY2 interacting protein 1, VCY2-interacting protein 1, VCY2IP1BPY2-interacting protein 1 | |
| MAP1S (AAH80547.1, 1 a.a. - 1059 a.a.) full-length human protein. MAAVAGSGAAAAPSSLLLVVGSEFGSPGLLTYVLEELERGIRSWDVDPGVCNLDEQLKVFVSRHSATFSSIVKGQRSLHHRGDNLETLVLLNPSDKSLYDELRNLLLDPASHKLLVLAGPCLEETGELLLQTGGFSPHHFLQVLKDREIRDILATTPPPVQPPILTITCPTFGDWAQLAPAVPGLQGALRLQLRLNPPAQLPNSEGLCEFLEYVAESLEPPSPFELLEPPTSGGFLRLGRPCCYIFPGGLGDAAFFAVNGFTVLVNGGSNPKSSFWKLVRHLDRVDAVLVTHPGADSLPGLNSLLRRKLAERSEVAAGGGSWDDRLRRLISPNLGVVFFNACEAASRLARGEDEAELALSLLAQLGITPLPLSRGPVPAKPTVLFEKMGVGRLDMYVLHPPSAGAERTLACVCALLVWHPAGPGEKVVRVLFPGCTPPACLLDGLVRLQHLRFLREPVVTPQDLEGPGRAESKESVGSRDSSKREGLLATHPRPGQERPGVARKEPARAEAPRKTEKEAKTPRELKKDPKPSVSRTQPREVRRAASSVPNLKKTNAQAAPKPRKAPSTSHSGFPPVANGPRSPPSLRCGEASPPSAACGSPASQLVATPSLELGPIPAGEEKALELPLAASSIPRPRTPSPESHRSPAEGSERLSLSPLRGGEAGPDASPTVTTPTVTTPSLPAEVGSPHSTEVDESLSVSFEQVLPPSAPTSEAGLSLPLRGPRARRSASPHDVDLCLVSPCEFEHRKAVPMAPAPASPGSSNDSSARSQERAGGLGAEETPPTSVSESLPTLSDSDPVPLAPGAADSDEDTEGFGVPRHDPLPDPLKVPPPLPDPSSICMVDPEMLPPKTARQTENVSRTRKPLARPNSRAAAPKATPVAAAKTKGLAGGDRASRPLSARSEPSEKGGRAPLSRKSSTPKTATRGPSGSASSRPGVSATPPKSPVYLDLAYLPSGSSAHLVDEEFFQRVRALCYVISGQDQRKEEGMRAVLDALLASKQHWDRDLQVTLIPTFDSVAMHTWYAETHARHQALGITVLGSNSMVSMQDDAFPACKVEF | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| AAH80547.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 55201 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction