missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2 Antibody (5H11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-92711CL1
This item is not returnable.
View return policy
Description
MAP2 Monoclonal antibody specifically detects MAP2 in Bovine,Human,Monkey,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry Free-Floating,Immunocytochemistry/Immunofluorescence
Specifications
| MAP2 | |
| Monoclonal | |
| CoraFluor 1 | |
| DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
| MAP2 Antibody (5H11) was developed against full length purified bovine protein, epitope mapped to projection domain of human sequence, amino acids 1057-1588 using MAP2 Projection Domain 3. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers | |
| 4133 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Free Floating), Immunocytochemistry/Immunofluorescence | |
| 5H11 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bovine, Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction