missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2 Antibody (5H11), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-92711V
This item is not returnable.
View return policy
Description
MAP2 Monoclonal specifically detects MAP2 in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.
Specifications
| MAP2 | |
| Monoclonal | |
| 1mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating | |
| DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
| Mouse | |
| 280 kDa | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers | |
| 4133 | |
| Human, Mouse, Rat, Bovine, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) | |
| 5H11 | |
| DyLight 405 | |
| P11137 | |
| MAP2 | |
| MAP2 Antibody (5H11) was developed against full length purified bovine protein, epitope mapped to projection domain of human sequence, amino acids 1057-1588 using MAP2 Projection Domain 3. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Note that since the epitope for this antibody in within the projection domain found only in MAP2A and MAP2B, and so the antibody does not bind to the lower molecular weight MAP2C and MAP2D isoforms which lack this region. | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction