missing translation for 'onlineSavingsMsg'
Learn More

MAP2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP3-05552AF405

Product Code. 18570020

  • € 679.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MAP2 Polyclonal antibody specifically detects MAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAP2
Polyclonal
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2
MAP2
0.1 mL
Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers
4133
Store at 4C in the dark.
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry
Alexa Fluor 405
50mM Sodium Borate
Goat
Immunogen affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.