missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2 Antibody (HM-2), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-11267AF405
This item is not returnable.
View return policy
Description
MAP2 Monoclonal antibody specifically detects MAP2 in Human, Mouse, Rat, Avian, Bovine, Chicken samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| MAP2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| P15146 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Avian, Bovine | |
| Purified |
| Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| HM-2 | |
| 50mM Sodium Borate | |
| DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
| This MAP2 antibody was developed against rat brain microtubule associated proteins (MAPs). | |
| 0.05 mL | |
| Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers | |
| 4133 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction