missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2K1 Monoclonal Antibody (OTI2F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
€ 242.00 - € 540.87
Specifications
| Antigen | MAP2K1 |
|---|---|
| Clone | OTI2F1 |
| Concentration | 1 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17515242
|
Origene Technologies
TA506007S |
30 μL |
€ 242.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17595232
|
Origene Technologies
TA506007 |
100 μL |
€ 540.87
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAP2K1 Monoclonal antibody specifically detects MAP2K1 in Canine, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| MAP2K1 | |
| 1 mg/mL | |
| Unconjugated | |
| Mouse | |
| Canine, Human, Mouse, Rat | |
| 170851, 26395, 478347, 5604 | |
| Primary |
| OTI2F1 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P31938, Q01986, Q02750 | |
| IgG2b | |
| -20° C, Avoid Freeze/Thaw Cycles |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel