missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2K3 Monoclonal Antibody (OTI1D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505631
This item is not returnable.
View return policy
Description
This gene encodes a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases, act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis.Specifications
| MAP2K3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O09110, P46734 | |
| MAP2K3 | |
| Full length human recombit protein of human MAP2K3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1D2 | |
| Unconjugated | |
| MAP2K3 | |
| MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 26397, 303200, 489547, 5606 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction