missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2K3 Monoclonal Antibody (OTI5G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505844
This item is not returnable.
View return policy
Description
This gene encodes a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases, act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis.Specifications
| MAP2K3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O09110, P46734 | |
| MAP2K3 | |
| Full length human recombit protein of human MAP2K3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI5G5 | |
| Unconjugated | |
| MAP2K3 | |
| MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 26397, 489547, 5606 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering