missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2K3 Monoclonal Antibody (OTI5G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505844S
This item is not returnable.
View return policy
Description
MAP2K3 Monoclonal antibody specifically detects MAP2K3 in Canine, Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| MAP2K3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P46734 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 489547, 5606 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI5G5 | |
| Unconjugated | |
| MAP2K3 | |
| AW212142; dual specificity mitogen-activated protein kinase kinase 3; EC 2.7.12.2; MAP kinase kinase 3; Map2k3; MAPK/ERK kinase 3; MAPKK 3; MAPKK3; MEK 3; Mek3; mitogen activated protein kinase kinase 3; mitogen-activated protein kinase kinase 3; Mkk3; mMKK3b; Prkmk3; protein kinase, mitogen-activated, kinase 3; SAPK kinase 2; SAPKK2; SAPKK-2; SKK2; Stress-activated protein kinase kinase 2 | |
| Full length human recombinant protein of human MAP2K3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction