missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP3K13 Monoclonal Antibody (OTI8A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812293S
This item is not returnable.
View return policy
Description
MAP3K13 Monoclonal antibody specifically detects MAP3K13 in Human samples. It is validated for Western BlotSpecifications
| MAP3K13 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96C23 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 130589 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8A4 | |
| Unconjugated | |
| GALM | |
| C130026N12Rik; Leucine zipper-bearing kinase; LZK; Map3k13; MEKK13; mitogen activated protein kinase kinase kinase 13; mitogen-activated protein kinase kinase kinase 13; Mixed lineage kinase; MLK | |
| Full length human recombinant protein of human GALM produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction