missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MAP3K19 (YSK4), GST Tag Recombinant Protein

Product Code. 15856591
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 15856591

Brand: Invitrogen™ A30501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This MAP3K19 (YSK4) recombinant human protein (941-end) was expressed in insect cells. MAP3K19 (YSK4) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic The gene accession number for MAP3K19 (YSK4) is NP_079328.3.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q56UN5
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 25% glycerol, 0.25mM DTT, 10mM glutathione, 150mM NaCl, 0.1mM EDTA, 0.1mM PMSF and no preservative; pH 7.5
Gene ID (Entrez) 80122
Molecular Weight (g/mol) 69.8 kDa
Name Human MAP3K19 (YSK4), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias MAP3K19; Mitogen-activated protein kinase kinase kinase 19; RCK; regulated in COPD kinase; regulated in COPD, protein kinase; SPS1/STE20-related protein kinase YSK4; yeast Sps1/Ste20-related kinase 4; YSK4; YSK4 Sps1/Ste20-related kinase homolog
Common Name MAP3K19
Gene Symbol MAP3K19
KinaseFamily MAP Kinases
Protein Length 941-end
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 941-end
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Active, Catalytic
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.