Learn More
MAP3K5 & APP Protein Protein Interaction Antibody Pair, Abnova™
Antibody Pair
Brand:  Abnova DI0061.1Set
Product Code. 16111094
  • € 677.00 / 1 set
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the MAP3K5 protein, and the other against the APP protein for use in in situ Proximity Ligation Assay.
Specifications
Specifications
MAP3K5
Unconjugated
ASK1/MAPKKK5/MEKK5
1 Set
Primary
Antibody pair set content: 1. MAP3K5 rabbit purified polyclonal antibody (20 ug) 2. APP mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
In situ PLA
mitogen-activated protein kinase kinase kinase 5
MAP3K5
RUO
4217/351
Human
Ab Pair