Learn More
MAP3K5 & APP Protein Protein Interaction Antibody Pair, Abnova™
Antibody Pair
Brand: Abnova DI0061.1Set
This item is not returnable.
View return policy
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the MAP3K5 protein, and the other against the APP protein for use in in situ Proximity Ligation Assay.Specifications
| MAP3K5 | |
| Unconjugated | |
| ASK1/MAPKKK5/MEKK5 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. MAP3K5 rabbit purified polyclonal antibody (20 ug) 2. APP mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| In situ PLA | |
| mitogen-activated protein kinase kinase kinase 5 | |
| MAP3K5 | |
| RUO | |
| 4217/351 | |
| Human | |
| Ab Pair |