missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP3K6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16212-1-AP-20UL
This item is not returnable.
View return policy
Description
Mitogen-activated protein kinase (MAPK) signaling cascades include MAPK or extracellular signal-regulated kinase (ERK), MAPK kinase (MKK or MEK), and MAPK kinase kinase (MAPKKK or MEKK). MAPKK kinase/MEKK phosphorylates and activates its downstream protein kinase, MAPK kinase/MEK, which in turn activates MAPK. The kinases of these signaling cascades are highly conserved, and homologs exist in yeast, Drosophila, and mammalian cells. MEKK6 activates the JNK, but not ERK or p38 kinase pathways.Specifications
| MAP3K6 | |
| Polyclonal | |
| Unconjugated | |
| MAP3K6 | |
| ASK2, MAP3K6, MAPKKK6, MEKK6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 313022, 53608, 9064 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95382, Q9WTR2 | |
| Map3k6 | |
| MAP3K6 Fusion Protein Ag9206 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction