missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP3K8/Tpl2/COT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 386.00 - € 500.00
Specifications
| Antigen | MAP3K8/Tpl2/COT |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18413321
|
Novus Biologicals
NBP1-86569-25ul |
25 μL |
€ 386.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18214677
|
Novus Biologicals
NBP1-86569 |
0.1 mL |
€ 500.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAP3K8/Tpl2/COT Polyclonal specifically detects MAP3K8/Tpl2/COT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MAP3K8/Tpl2/COT | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Cancer, Protein Kinase | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 1326 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ENLYASEEPAVYEPSLMTMCQDSNQNDERSKSLLLSGQEVPWLSSVRYGTVEDLLAFANHISNTAKHFYGQRPQESGILLNMVITPQNGRYQIDSDVLLIPWKLTYRNIGSDFIPRGAFGKVYLAQDIKTKKRMACKLIPVDQFKPSDV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Unconjugated | |
| RUO | |
| Human | |
| Cancer Osaka thyroid oncogene, c-COT, cot (cancer Osaka thyroid) oncogene, COTMEKK8, EC 2.7.11.25, ESTF, ESTFLJ10486, Ewing sarcoma transformant, mitogen-activated protein kinase kinase kinase 8, Proto-oncogene c-Cot, proto-oncogene serine/threoine protein kinase, Serine/threonine-protein kinase cot, Tpl-2, TPL2, Tumor progression locus 2, tumor progression locus-2 | |
| MAP3K8 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title