missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP4K3 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99100FR
This item is not returnable.
View return policy
Description
MAP4K3 Polyclonal antibody specifically detects MAP4K3 in Human samples. It is validated for Western Blot
Specifications
| MAP4K3 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 680 | |
| EC 2.7.11, EC 2.7.11.1, germinal center kinase-like kinase, Germinal center kinase-related protein kinase, GLKMEKKK 3, MAPK/ERK kinase kinase kinase 3, MAPKKKK3, MEK kinase kinase 3, mitogen-activated protein kinase kinase kinase kinase 3, RAB8IPL1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MAP4K3. | |
| 0.1 mL | |
| Lipid and Metabolism, Protein Kinase | |
| 8491 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction