Learn More
Novus Biologicals™ MAP6D1 Antibody (Genentech patent anti-STOP-1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-27974-50ug
 View more versions of this product
Product Code. 30047207
  • € 213.00 / 50µg
Description
MAP6D1 Monoclonal antibody specifically detects MAP6D1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MAP6D1
Monoclonal
Unconjugated
Q9C8N5
Human
Protein A purified
RUO
79929
Human
Purified
Flow Cytometry, ELISA, Functional Assay
Genentech patent anti-STOP-1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
FLJ12748, MAP6 domain containing 1, MAP6 domain-containing protein 1,21 kDa STOP-like protein, MAPO6D1, SL21, STOP-like protein 21
STOP1
50 μg
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.