missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP7D2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563744
This item is not returnable.
View return policy
Description
Immunogen sequence: PLNPSYKSSP TRNIEKKKAT STSTSGAGDV GKEALSGGEA SLVEKVKRGQ RTATSLPVVN FGSPLRRCEF SGGIPKRPSS PVI Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 60%.
Specifications
| MAP7D2 | |
| Polyclonal | |
| Unconjugated | |
| MAP7D2 | |
| MAP7 domain containing 2; MAP7 domain containing 2 brain; MAP7 domain containing 2 protein varaint 1; MAP7 domain containing 2 protein varaint 2; MAP7 domain-containing protein 2; MAP7 domain-containing protein 2 brain specific variant 1; MAP7 domain-containing protein 2 brain specific variant 2; MAP7D2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 256714 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96T17 | |
| MAP7D2 | |
| Recombinant protein corresponding to Human MAP7D2. Recombinant protein control fragment (Product #RP-104206). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction