Learn More
Mapenterol Hydrochloride, TRC
Product Code. 17952485
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952485 25 mg 25mg
17982125 10 mg 10mg
17903495 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17952485 Supplier Toronto Research Chemicals Supplier No. TRCM18700025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54238-51-6
Molecular Formula C14 H20 Cl F3 N2 O . Cl H
Molecular Weight (g/mol) 361.23
Synonym Benzenemethanol, 4-amino-3-chloro-α-[[(1,1-dimethylpropyl)amino]methyl]-5-(trifluoromethyl)-, hydrochloride (1:1), Benzenemethanol, 4-amino-3-chloro-α-[[(1,1-dimethylpropyl)amino]methyl]-5-(trifluoromethyl)-, monohydrochloride (9CI), 4-Amino-3-chloro-α-[[(1,1-dimethylpropyl)amino]methyl]-5-(trifluoromethyl)benzenemethanol hydrochloride (1:1), Mapenterol hydrochloride
IUPAC Name 1-[4-amino-3-chloro-5-(trifluoromethyl)phenyl]-2-(2-methylbutan-2-ylamino)ethanol;hydrochloride
SMILES Cl.CCC(C)(C)NCC(O)c1cc(Cl)c(N)c(c1)C(F)(F)F
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C14H20ClF3N2O.ClH/c1-4-13(2,3)20-7-11(21)8-5-9(14(16,17)18)12(19)10(15)6-8;/h5-6,11,20-21H,4,7,19H2,1-3H3;1H
Formula Weight 360.0983
Percent Purity >95%
Chemical Name or Material Mapenterol hydrochloride
Grade HPLC