missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK1, Rabbit anti-Human, FITC, Clone: A11, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB18990.10Reactions
This item is not returnable.
View return policy
Specifications
| MAPK1 | |
| Monoclonal | |
| FITC | |
| In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide). | |
| ERK/ERK2/ERT1/MAPK2/P42MAPK/PRKM1/PRKM2/p38/p40/p41/p41mapk | |
| Rabbit | |
| 10 Reactions | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| A11 | |
| Flow Cytometry (5 uL/106 cells or 0.05 ug/mL) The optimal working dilution should be determined by the end user. | |
| mitogen-activated protein kinase 1 | |
| MAPK1 | |
| A synthetic phosphopeptide corresponding to residues surrounding T202/Y204 of human MAPK1/MAPK3. | |
| RUO | |
| 5594/5595 | |
| Store at 2-8°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction