missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK13 Monoclonal Antibody (OTI2A10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505856S
This item is not returnable.
View return policy
Description
MAPK13 Monoclonal antibody specifically detects MAPK13 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| MAPK13 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15264 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5603 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A10 | |
| Unconjugated | |
| MAPK13 | |
| im:7136778; MAP kinase 13; MAP kinase p38 delta; MAPK 13; Mapk13; MAPK-13; mitogen activated protein kinase 13; mitogen-activated protein kinase 13; Mitogen-activated protein kinase p38 delta; p38 delta MAP kinase; p38delta; PRKM13; SAPK/Erk/kinase 4; SAPK4; Serk4; Stress-activated protein kinase 4 | |
| Full length human recombinant protein of human MAPK13 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction