missing translation for 'onlineSavingsMsg'
Learn More

MAPK13 Monoclonal Antibody (OTI2C7), TrueMAB™, OriGene

Product Code. p-200011636
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17534922 30 μL 30µL
17514922 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17534922 Supplier Origene Technologies Supplier No. TA505866S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAPK13 Monoclonal antibody specifically detects MAPK13 in Canine, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen MAPK13
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MAPK13
Gene Accession No. O15264, Q9WTY9, Q9Z1B7
Gene Alias im:7136778; MAP kinase 13; MAP kinase p38 delta; MAPK 13; Mapk13; MAPK-13; mitogen activated protein kinase 13; mitogen-activated protein kinase 13; Mitogen-activated protein kinase p38 delta; p38 delta MAP kinase; p38delta; PRKM13; SAPK/Erk/kinase 4; SAPK4; Serk4; Stress-activated protein kinase 4
Host Species Mouse
Immunogen Full length human recombinant protein of human MAPK13 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26415, 29513, 5603, 612821
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.