Learn More
MAPK13 Monoclonal Antibody (OTI2G6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505865S
Encompass_Preferred
 View more versions of this product
Product Code. 17504922
  • € 242.00 / 30µL
Description
MAPK13 Monoclonal antibody specifically detects MAPK13 in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications
Specifications
MAPK13
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9H2B2
Mouse
Affinity chromatography
RUO
6860
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunocytochemistry, Western Blot
OTI2G6
Unconjugated
Syt4
im:7136778; MAP kinase 13; MAP kinase p38 delta; MAPK 13; Mapk13; MAPK-13; mitogen activated protein kinase 13; mitogen-activated protein kinase 13; Mitogen-activated protein kinase p38 delta; p38 delta MAP kinase; p38delta; PRKM13; SAPK/Erk/kinase 4; SAPK4; Serk4; Stress-activated protein kinase 4
Full length human recombinant protein of human SYT4 produced in HEK293T cell.