missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK13, Mouse anti-Human, Clone: 2B2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005603-M05.100ug
This item is not returnable.
View return policy
Description
Sequence: NDKAAKSYIQSLPQTPRKDFTQLFPRASPQAADLLEKMLELDVDRRLTAAQALTHPFFEPFRDPEEETEAQQPFDDSLEHEKLTVDEWKQHIYKEIVNFSPIARKDSRRRSGMKLSpecifications
| MAPK13 | |
| Monoclonal | |
| Unconjugated | |
| mitogen-activated protein kinase 13 | |
| MGC99536/PRKM13/SAPK4/p38delta | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2B2 | |
| In 1x PBS, pH 7.4 | |
| BC000433 | |
| MAPK13 | |
| MAPK13 (AAH00433, 251 a.a. ∼ 365 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5603 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction