missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MAPK1IP1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29351
This item is not returnable.
View return policy
Description
MAPK1IP1L Polyclonal antibody specifically detects MAPK1IP1L in Human samples. It is validated for Immunoprecipitation
Specifications
| MAPK1IP1L | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 93487 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| c14_5346, C14orf32, chromosome 14 open reading frame 32, MAPK-interacting and spindle-stabilizing protein, MAPK-interacting and spindle-stabilizing protein-like, MGC23138, MISS, mitogen activated protein kinase 1 interacting protein 1-like, mitogen-activated protein kinase 1 interacting protein 1-like, Mitogen-activated protein kinase 1-interacting protein 1-like | |
| Between 1 and 50 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction