missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK3, Rabbit anti-Human, Clone: G15-B, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB15896.100uL
This item is not returnable.
View return policy
Specifications
| MAPK3 | |
| Monoclonal | |
| Unconjugated | |
| In 20 mM Tris-HCl buffer, pH 8.0 (10 mg/mL BSA, 0.05% Sodium Azide). | |
| ERK1/HS44KDAP/HUMKER1A/MGC20180/P44ERK1/P44MAPK/PRKM3 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| G15-B | |
| ELISA (1:20000-1:100000) Immunocytochemistry Immunoprecipitation Western Blot (1:5000) The optimal working dilution should be determined by the end user. | |
| mitogen-activated protein kinase 3 | |
| MAPK3 | |
| A synthetic peptide corresponding to the protein area including conserved pT-E-pY motif of activated MAPK3/MAPK1. | |
| RUO | |
| 5595/5594 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction