missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK8, Mouse anti-Human, Clone: 4H5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005599-M05.100ug
This item is not returnable.
View return policy
Description
Sequence: HPYINVWYDPSEAEAPPPKIPDKQLDEREHTIEEWKELIYKEVMDLEERTKNGVIRGQPSPLGAAVINGSQHPSSSSSVNDVSSMSTDPTLASDTDSSLEAAAGPLGCCRSpecifications
| MAPK8 | |
| Monoclonal | |
| Unconjugated | |
| mitogen-activated protein kinase 8 | |
| JNK/JNK1/JNK1A2/JNK21B1/2/PRKM8/SAPK1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunoprecipitation, Western Blot | |
| 4H5 | |
| In 1x PBS, pH 7.4 | |
| NM_139049 | |
| MAPK8 | |
| MAPK8 (NP_620637, 318 a.a. ∼ 427 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5599 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction