missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK8IP1 Monoclonal Antibody (OTI13F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809926S
This item is not returnable.
View return policy
Description
MAPK8IP1 Monoclonal antibody specifically detects MAPK8IP1 in Human samples. It is validated for Western BlotSpecifications
| MAPK8IP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y2H2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 22876 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI13F10 | |
| Unconjugated | |
| INPP5F | |
| C-Jun-amino-terminal kinase-interacting protein 1; Ib1; IB-1; Islet-brain 1; islet-brain-1; JIP1; JIP-1; JIP-1-related protein; JNK MAP kinase scaffold protein 1; JNK-interacting protein 1; JRP; Mapk8ip; Mapk8ip1; mitogen activated protein kinase 8 interacting protein; mitogen activated protein kinase 8 interacting protein 1; mitogen-activated protein kinase 8 interacting protein 1; mitogen-activated protein kinase 8-interacting protein 1; mjip-2a; PRKM8 interacting protein; PRKM8IP; protein kinase, mitogen-activated 8 interacting protein; Skip | |
| Full length human recombinant protein of human INPP5F produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction