missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPKAP Kinase 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99449-100ul
This item is not returnable.
View return policy
Description
MAPKAP Kinase 3 Polyclonal antibody specifically detects MAPKAP Kinase 3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MAPKAP Kinase 3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| 3PKEC 2.7.11.1, Chromosome 3p kinase, EC 2.7.11, MAP kinase-activated protein kinase 3, MAPK-activated protein kinase 3, MAPKAP kinase 3, MAPKAP33pK, MAPKAPK-3, mitogen-activated protein kinase-activated protein kinase 3 | |
| Produced in rabbits immunized with purified, recombinant Human MAPKAP Kinase 3 (Accession#: NP_004626.1; Met1-Gln382) | |
| 100 μg | |
| Protein Kinase | |
| 7867 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction