missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLOD4 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18449B
This item is not returnable.
View return policy
Description
GLOD4 Polyclonal antibody specifically detects GLOD4 in Human, Mouse samples. It is validated for Western Blot
Specifications
| GLOD4 | |
| Polyclonal | |
| Western Blot | |
| C17orf25, CGI-150, chromosome 17 open reading frame 25, glyoxalase domain containing 4, glyoxalase domain-containing protein 4, HC71 | |
| Recognizes region between residues Between Between 263 and 313.. | |
| 0.1 mL | |
| Cancer | |
| 51031 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction