missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPKAPK5 Monoclonal Antibody (OTI4G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804232S
This item is not returnable.
View return policy
Description
MAPKAPK5 Monoclonal antibody specifically detects MAPKAPK5 in Human samples. It is validated for Western BlotSpecifications
| MAPKAPK5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IW41 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8550 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI4G4 | |
| Unconjugated | |
| MAPKAPK5 | |
| MAP kinase-activated protein kinase 5; MAPK-activated protein kinase 5; MAPKAP kinase 5; Mapkapk5; MAPKAP-K5; MAPKAPK-5; mitogen-activated protein kinase-activated protein kinase 5; MK5; MK-5; p38-regulated/activated protein kinase; PRAK | |
| Human recombinant protein fragment corresponding to amino acids 197-471 of human MAPKAPK produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction