Learn More
MAPKAPK5 Mouse anti-Human, Clone: OTI1B4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF804189
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the serine/threonine kinase family. In response to cellular stress and proinflammatory cytokines, this kinase is activated through its phosphorylation by MAP kinases including MAPK1/ERK, MAPK14/p38-alpha, and MAPK11/p38-beta. In vitro, this kinase phosphorylates heat shock protein HSP27 at its physiologically relevant sites. Two alternately spliced transcript variants of this gene encoding distinct isoforms have been reported.Specifications
| MAPKAPK5 | |
| Monoclonal | |
| Unconjugated | |
| MAPKAPK5 | |
| MAPKAP-K5, MK-5, MK5, PRAK | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8550 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1B4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8IW41 | |
| MAPKAPK5 | |
| Human recombit protein fragment corresponding to amino acids 197-471 of human MAPKAPK5 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.