Learn More
MAPT (phospho S262) Rabbit anti-Human, Mouse, Rabbit, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MAPT.
Brand: Abnova PAB29609.100uL
Description
This gene encodes the microtubule-associated protein tau (MAPT) whose transcript undergoes complex, regulated alternative splicing, giving rise to several mRNA species. MAPT transcripts are differentially expressed in the nervous system, depending on stage of neuronal maturation and neuron type. MAPT gene mutations have been associated with several neurodegenerative disorders such as Alzheimer's disease, Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy. [provided by RefSeq]
Specifications
| MAPT (phospho S262) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MAPT. | |
| In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| MAPT | |
| A synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S262 of human MAPT. | |
| 100 μL | |
| Primary | |
| MAPT (phospho S262) polyclonal antibody detects endogenous level of MAPT only when phosphorylated at serine 262. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunofluorescence (1:100-1:200) Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| DDPAC/FLJ31424/FTDP-17/MAPTL/MGC138549/MSTD/MTBT1/MTBT2/PPND/TAU | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4137 | |
| Human, Mouse, Rabbit | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.