missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Marapsin/Pancreasin Antibody (MM0463-3H15), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11786AF488
This item is not returnable.
View return policy
Description
Marapsin/Pancreasin Monoclonal specifically detects Marapsin/Pancreasin in Human samples. It is validated for Western Blot.
Specifications
| Marapsin/Pancreasin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PRSS27 | |
| Human recombinant Marapsin/Pancreasin | |
| 0.1 mL | |
| 83886 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| MM0463-3H15 | |
| Western Blot | |
| CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPNMarapsin, Pancreasin, protease, serine 27, serine protease 27 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction