missing translation for 'onlineSavingsMsg'
Learn More

Marbon CNB 23010, TRC

Product Code. 30474734
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474734 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474734 Supplier Toronto Research Chemicals Supplier No. TRCM19720010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Marbon CNB 23010
CAS 26595-57-3
Molecular Formula C4 H5 N3 O2
Quantity 10 mg
Synonym Marbon CNB 23010
InChI Formula InChI=1S/C4H5N3O2/c5-4-6-2(8)1-3(9)7-4/h1H,(H4,5,6,7,8,9)
SMILES Nc1nc(O)cc(O)n1
IUPAC Name 2-aminopyrimidine-4,6-diol
Molecular Weight (g/mol) 127.101
Formula Weight 127.038
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.