missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MARCH11 Polyclonal Antibody

Product Code. 15817653
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15817653 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15817653 Supplier Invitrogen™ Supplier No. PA569239

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MARCH11 Polyclonal Antibody for Western Blot

E3 ubiquitin-protein ligase that mediates polyubiquitination of CD4. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates. May play a role in ubuquitin-dependent protein sorting in developmenting spermatids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MARCH11
Gene Accession No. A6NNE9
Gene Alias 11-Mar; 9630025C22; E3 ubiquitin-protein ligase MARCH11; MARCH11; MARCH-XI; membrane associated ring finger 11; membrane associated ring-CH-type finger 11; membrane-associated ring finger (C3HC4) 11; membrane-associated RING finger protein 11; Membrane-associated RING-CH protein XI; RGD1559945; RING-type E3 ubiquitin transferase MARCH11
Gene Symbols MARCH11
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human MARCH11.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441061
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.