missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCH8 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76959PCP
This item is not returnable.
View return policy
Description
MARCH8 Polyclonal antibody specifically detects MARCH8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| MARCH8 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| Cellular modulator of immune recognition, EC 6.3.2, EC 6.3.2.-, MARCH-VIIIcellular modulator of immune recognition, membrane-associated ring finger (C3HC4) 8, Membrane-associated RING finger protein 8, Membrane-associated RING-CH protein VIII, MIR, RING finger protein 178 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human MARCH8. The immunogen is located within the last 50 amino acids of MARCH8. Amino Acid Squence: CPETSKKNIFEKSPLTE | |
| 0.1 mL | |
| Zinc Finger | |
| 220972 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction