missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCKS Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
€ 321.00 - € 481.00
Specifications
| Antigen | MARCKS |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:2000, Immunocytochemistry/ Immunofluorescence 1:2000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642907
|
Novus Biologicals
NBP3-05539-100ul |
100 μg |
€ 481.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690509
|
Novus Biologicals
NBP3-05539-50ul |
50 μg |
€ 321.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MARCKS Monoclonal antibody specifically detects MARCKS in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| MARCKS | |
| Western Blot 1:2000, Immunohistochemistry 1:2000, Immunocytochemistry/ Immunofluorescence 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Monkey | |
| 80K-L, 80K-L protein, FLJ14368, MACSmyristoylated alanine-rich C-kinase substrate, MRACKS, myristoylated alanine-rich protein kinase C substrate, myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L), phosphomyristin, PKCSLFLJ90045, PRKCSL, Protein kinase C substrate, 80 kDa protein, light chain | |
| Full length human MARCKS expressed in and purified from E. coli. | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism, Signal Transduction | |
| 50% PBS, 50% glycerol | |
| 4082 | |
| IgG1 | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title