missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCKS Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP3-05540-50ul
This item is not returnable.
View return policy
Description
MARCKS Polyclonal antibody specifically detects MARCKS in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| MARCKS | |
| Polyclonal | |
| Western Blot 1:5000-1:10000, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/ Immunofluorescence 1:500-1:1000 | |
| 80K-L, 80K-L protein, FLJ14368, MACSmyristoylated alanine-rich C-kinase substrate, MRACKS, myristoylated alanine-rich protein kinase C substrate, myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L), phosphomyristin, PKCSLFLJ90045, PRKCSL, Protein kinase C substrate, 80 kDa protein, light chain | |
| Full length recombinant human MARCKS expressed in and purified from E. coli. | |
| 50 μg | |
| Lipid and Metabolism, Signal Transduction | |
| 4082 | |
| Store at 4°C. | |
| IgY |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Supplied as an aliquot of IgY preparation | |
| Chicken | |
| IgY purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction