missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCO Antibody (579511), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2956D
This item is not returnable.
View return policy
Description
MARCO Monoclonal antibody specifically detects MARCO in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| MARCO | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 579511 | |
| Flow Cytometry, CyTOF-ready | |
| macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse MARCO, Gln70-Ser518, Accession # Q60754 | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 8685 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction