Learn More
Marginal Zone B Cells Mouse anti-Rat, BB700, Clone: HIS57, BD OptiBuild™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 746119
Product Code. 15960290
  • ONLINE EXCLUSIVE PRICE
    € 389.00
    € 388.12 / Each
    Save € 0.88
    0.23% Off
Description
Marginal Zone B Cells Monoclonal antibody specifically detects Marginal Zone B Cells in Rat samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Marginal Zone B Cells
Monoclonal
0.2 mg/mL
Mouse
RUO
Rat
Purified
Flow Cytometry
HIS57
Brilliant Blue 700 (BB700)
50 μg
Primary
Refrigerate (2 to 8°C)
IgG1 κ