missing translation for 'onlineSavingsMsg'
Learn More

Maribavir, MedChemExpress

Product Code. 16489567
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489567 5 mg 5mg
16443077 50 mg 50mg
16437086 100 mg 100mg
16427028 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489567 Supplier Medchem Express Supplier No. HY16305

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Maribavir is a potent inhibitor of histone phosphorylation catalyzed by wild-type pUL97 in vitro, with an IC50 of 3 nM. Maribavir has potent antiviral activity against HCMV and Epstein-Barr virus (EBV).

Maribavir is a potent inhibitor of histone phosphorylation catalyzed by wild-type pUL97 in vitro, with an IC50 of 3 nM. Maribavir has potent antiviral activity against HCMV and Epstein-Barr virus (EBV).

Specifications

Chemical Name or Material Maribavir
CAS 176161-24-3
Color White
Purity Grade Notes Research
Molecular Formula C15H19Cl2N3O4
Quantity 5 mg
Synonym 1263W94 BW1263W94 GW257406X
Solubility Information DMSO : 200 mg/mL (531.58 mM; Need ultrasonic)
SMILES ClC1=C(Cl)C=C2C(N=C(NC(C)C)N2[C@@H]3[C@@H](O)[C@@H](O)[C@H](CO)O3)=C1
Molecular Weight (g/mol) 376.24
Formula Weight 376.24
Percent Purity 98.16%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.